Orders LinkContactUsLinkComponent
Ammoniumchloride
Ammoniumchloride
5 kg
Catalog # ROTH5470.2
Irritant
Marketsource
Supplier:   Roth Carl
Ammoniumchloride
Catalog # ROTH5470.2
Supplier:  Roth Carl
Marketsource
CAS Number:  12125-02-9
Ammoniumchloride
Ammoniumchloride
5 kg
CAS Number:  12125-02-9
Price
Inquire For Price
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Ammonium muriate, Salmiac
  • CAS Number:12125-02-9
  • Formula:NH₄Cl
  • MDL Number:MFCD00011420
  • Molecular Weight:53.49 g/mol
  • Merck Index:13,00510
  • Melting Pt:335 °C
  • Density:1,5256 g/cm³ (20 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:235-186-4
Specifications
Specifications
  • VPE:
    5 kg
Specifications