Ethidiumbromide 10 mg/ml in aqueous solution
Glass bottle, 30 mL
Catalogus # ROTH2218.2
Marketsource
Leverancier: Roth Carl
Ethidiumbromide 10 mg/ml in aqueous solution
Catalogus # ROTH2218.2
Leverancier: Roth Carl
Marketsource
CAS-nummer: 1239-45-8
Ga naar:
Product Details & Documents
Synoniemen: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
- CAS-nummer:1239-45-8
- Formule:C₂₁H₂₀BrN₃
- MDL-nummer:MFCD00011724
- Molecular Weight:394.31 g/mol
- EINECS:214-984-6
Documents
Certificaten
Zoeken op lotnummerSpecificaties
Specificaties
- Verpakking:Glass bottle
- VPE:30 mL



