Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
NS-stoppen
NS-stoppen
14/23, Flat
  217-9093
NS-stoppen
  217-9093
NS-stoppen
NS-stoppen
14/23, Flat
4,20 €
1 * 1 ST
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certifications: DIN 12252
Ordering information: Available with flat or pointed bottom.
Documents
  • (NS):
    14/23
  • Type:
    Flat
  • Seq:
    13