Orders LinkContactUsLinkComponent
Cold traps, Type KF
Cold traps, Type KF
Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mm
Catalogus # KGWA17402
Leverancier:   KGW
Cold traps, Type KF
Catalogus # KGWA17402
Leverancier:  KGW
Cold traps, Type KF
Cold traps, Type KF
Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mm
Price
747,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminium
Diensten: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
  • Omschrijving:
    Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mm
  • Seq:
    07
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Plastic ring for cold trap, type KF
    Plastic ring for cold trap, type KF
    KGWA1733
    Log in voor beschikbaarheid
    1 * 1 ST - 62,50 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Cooling finger S 29-GL
    Cooling finger S 29-GL
    KGWA1741
    Log in voor beschikbaarheid
    1 * 1 ST - 275,00 €