Molecular models: RNA, Molymod®
Molecular model of mRNA
Catalogus # SPIRMRNA-24-PD
Marketsource
Leverancier: SPIRING ENTERPRISES
Molecular models: RNA, Molymod®
Catalogus # SPIRMRNA-24-PD
Leverancier: SPIRING ENTERPRISES
Marketsource
Ga naar:
Product Details & Documents
The Molymod system is the original dual-scale system of high quality molecular models.
Levering: Protein synthesis kit with: 24 bases, 12 ribose/phophate, 4 TRNA, 4 amino acid parts.
Specificaties
- Omschrijving:Molecular model of mRNA
- Seq:002
- Cat. No.:SPIRMRNA-24-PD