Orders LinkContactUsLinkComponent
Capillary column caps
Capillary column caps
Capillary column caps, glass
Catalogus # REST21044
Leverancier:   Restek
Capillary column caps
Catalogus # REST21044
Leverancier:  Restek
Capillary column caps
Capillary column caps
Capillary column caps, glass
Price
142,00 €
Log in voor beschikbaarheid
1 * 20 ST
Product Details & Documents
These non-reusable column caps increase column lifetime, prevent moisture and air from entering the column during storage.
  • Attach to the column in seconds to form an airtight seal
  • Available in two styles, glass or silicone material
  • Glass caps are colour coded for identifying detector and injector ends
Specificaties
  • Omschrijving:
    Capillary column caps, glass
  • Seq:
    01