Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux
Fractionating columns, Vigreux
300 mm, 29/32 NS, 29/32 NS
Catalogus # 201-0178
Leverancier:   witeg Labortechnik
Fractionating columns, Vigreux
Catalogus # 201-0178
Leverancier:  witeg Labortechnik
Fractionating columns, Vigreux
Fractionating columns, Vigreux
300 mm, 29/32 NS, 29/32 NS
Price
106,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.
Specificaties
  • Afmetingen kern:
    29/32 NS
  • Huls:
    29/32 NS
  • Totale lengte:
    300 mm