Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux
Fractionating columns, Vigreux
300 mm, 24/29 NS, 24/29 NS
Catalog # 201-0177
Supplier:   witeg Labortechnik
Fractionating columns, Vigreux
Catalog # 201-0177
Supplier:  witeg Labortechnik
Fractionating columns, Vigreux
Fractionating columns, Vigreux
300 mm, 24/29 NS, 24/29 NS
Price
106,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.
Specifications
  • Afmetingen kern:
    24/29 NS
  • Huls:
    24/29 NS
  • Totale lengte:
    300 mm