Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux
Fractionating columns, Vigreux
200 mm, 19/26 NS, 19/26 NS
Catalogus # 201-0175
Leverancier:   witeg Labortechnik
Fractionating columns, Vigreux
Catalogus # 201-0175
Leverancier:  witeg Labortechnik
Fractionating columns, Vigreux
Fractionating columns, Vigreux
200 mm, 19/26 NS, 19/26 NS
Price
87,30 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.
Specificaties
  • Afmetingen kern:
    19/26 NS
  • Huls:
    19/26 NS
  • Totale lengte:
    200 mm