Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Glass wool
Glass wool
Base-deactivated glass wool, ideal for amines and other basic compounds
Catalogus # REST20999
Leverancier:   Restek
Glass wool
Catalogus # REST20999
Leverancier:  Restek
Glass wool
Glass wool
Base-deactivated glass wool, ideal for amines and other basic compounds
Price
81,40 €
Log in voor beschikbaarheid
1 * 10 g
Product Details & Documents
Specificaties
  • Omschrijving:
    Base-deactivated glass wool, ideal for amines and other basic compounds
  • Seq:
    03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.