Orders LinkContactUsLinkComponent
Glass wool
Glass wool
Deactivated borosilicate glass wool, more flexible than fused silica wool
Catalog # REST20789
Supplier:   Restek
Glass wool
Catalog # REST20789
Supplier:  Restek
Glass wool
Glass wool
Deactivated borosilicate glass wool, more flexible than fused silica wool
Price
51,50 €
Log in for availability
1 * 10 g
Product Details & Documents
Specifications
  • Omschrijving:
    Deactivated borosilicate glass wool, more flexible than fused silica wool
  • Seq:
    02