Order Entry
Belgium
ContactUsLinkComponent
CELLMASTER# roller bottles 2,5X, van heldere, breukvaste kunststof (PET) non pyrogenic , DNase/RNase free, oppervl. 2125 cm, vu lvolume: 2500 ml, graduering 50-1000 ml, lengte 275 mm, diam. 122 mm, per 5 onderverpakt STERIEL 1 * 20 ST
Catalogus # 392-0068
Leverancier:  Greiner Bio-One
CELLMASTER# roller bottles 2,5X, van heldere, breukvaste kunststof (PET) non pyrogenic , DNase/RNase free, oppervl. 2125 cm, vu lvolume: 2500 ml, graduering 50-1000 ml, lengte 275 mm, diam. 122 mm, per 5 onderverpakt STERIEL 1 * 20 ST
Catalogus # 392-0068
Leverancier:  Greiner Bio-One
Frequently Bought Together