Pipette tips
VistaTip™, pyrogen-free, 200 µl, +, Individually wrapped
N° de catalogue VIST2027
Fournisseur: VISTALAB TECHNOLOGIES
Aller à :
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Spécifications
- Verpakt:Individually wrapped
- Steriel:+
- Volume:200 µl
- Seq:02
- Tip stijl:VistaTip™, pyrogen-free