Pycnometers, unadjusted
2 ml
Catalogus # WITG3.900.002
Leverancier: witeg Labortechnik
Ga naar:
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in capillary stopper
- Nominal capacity printed on the bottom
- Very good chemical and temperature resistance
Certificeringen: DIN 12797
Specificaties
- Volume (nom.):2 ml