Celtellers, Nageotte
Counting chamber, bright-lined, Nageotte, 0,5 mm
Catalog # MARI0640530
Supplier: MARIENFELD
Jump to:
Product Details & Documents
These counting chambers are designed for leucocytes and erythrocytes in in vitro applications.
Certifications: IVD directive 98/79/EC.
Specifications
- Kamerdiepte:0,5 mm
- Omschrijving:Counting chamber, bright-lined, Nageotte
- Seq:001
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.