Orders LinkContactUsLinkComponent
Celtellers, Nageotte
Celtellers, Nageotte
Counting chamber, bright-lined, Nageotte, 0,5 mm
Catalog # MARI0640530
Supplier:   MARIENFELD
Celtellers, Nageotte
Catalog # MARI0640530
Supplier:  MARIENFELD
Celtellers, Nageotte
Counting chamber, bright-lined, Nageotte, 0,5 mm
Price
158,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These counting chambers are designed for leucocytes and erythrocytes in in vitro applications.
Certifications: IVD directive 98/79/EC.
Specifications
  • Kamerdiepte:
    0,5 mm
  • Omschrijving:
    Counting chamber, bright-lined, Nageotte
  • Seq:
    001