Super syringes
Super syringe with TLL connector, 1,5 l, S1500 TLL
Catalogus # HAMI86313
Leverancier: HAMILTON BONADUZ
Ga naar:
Product Details & Documents
Super syringes with PTFE Luer lock (TLL) connector or tracheal adapter.
- Clear acrylic syringe barrels
- For use with gases and liquids
Ideal for use with air sampling, preparing gas standards, calibrating reservoirs, and pneumographs.
Specificaties
- Type:S1500 TLL
- Volume:1,5 l
- Omschrijving:Super syringe with TLL connector
- Seq:03
Accessories
Productinformatie
- Catalogus #VoorBeschikbaarheidPrice
- TLL connectorTLL connectorHAMI18026super syringe S0500, S1000, S1500, S2000Log in voor beschikbaarheid1 * 1 ST - 85,50 €