Slide holders, DrainRack™
Slide holders, wit, 200
Catalogus # 631-1913
Leverancier: Simport Scientific
Ga naar:
Product Details & Documents
This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individually numbered slots. All slides are stored upright for easier insertion and removal.
To save space, the number of slides held can be doubled simply by placing two slides in each slot, making room for 200 slides instead of 100.
Barcodes can be read without having to remove the slides from the box.
Specificaties
- Aantal objectglaasjes:200
- Kleur:wit
- Omschrijving:Slide holders