Caps for tubes
Cap for 11 mm tubes, rood
Catalogus # DELT305306
Leverancier: DELTALAB
Ga naar:
Product Details & Documents
Polyethylene caps for all types of polypropylene and polystyrene tubes.
Documents
Specificaties
- Kleur:rood
- Omschrijving:Cap for 11 mm tubes
- Seq:04
- Cat. No.:DELT305306