Voedselsondes
Lange sonde, Roestvrij staal, 3×200 mm
Catalog # 620-0175
Supplier: TESTO
Jump to:
Product Details & Documents
These temperature probes are suitable for measurements in either semi-solid or liquid media in the food sector.
For use with the food thermometer Testo 105.
Services: Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
Specifications
- Omschrijving:Lange sonde
- Materiaal:Roestvrij staal
- Ø×l:3×200 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.