Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Conjugate stabilisers

Conjugate stabilisers

ALP Conjugate Stabiliser, 100 mL
Catalogus # ROCKMB-061-0100
Leverancier:   Rockland Immunochemicals
Conjugate stabilisers
Catalogus # ROCKMB-061-0100
CAS-nummer:  
Conjugate stabilisers

ALP Conjugate Stabiliser, 100 mL

CAS-nummer:  
Price
184,00 €
Log in voor beschikbaarheid
1 * 100 mL
Product Details & Documents

Conjugate stabilisers are used with horseradish peroxidase (HRP) and calf intestinal alkaline phosphatase conjugated primary or secondary antibodies or other proteins or compounds in solution. The components of the stabiliser (proprietary) are unique and stabilise both the antibody and enzyme portions of the conjugate.

This conjugate stabiliser prevents the loss of catalytic activity and maintains the active conformation of the antibody portion of the conjugate. This 1X diluent buffer contains buffered saline along with proprietary reagents to stabilise and preserve both HRP and ALP conjugated primary or secondary antibodies in solution. A proprietary combination of preservatives is used that is azide- and mercury-free.

Delivery information: Supplied as ready to use sterile filtered liquid.

Specificaties

Specificaties

  • VPE:
    100 mL
  • Omschrijving:
    ALP Conjugate Stabiliser

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.