Xyleen ≥97%
Glass bottle, 1.000 mL
Purity: 97+% CAS No.: 1330-20-7 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 29°(84°F) Refractive Index: 1.4970 Odour: Aromatic Melting Point: -25° Boiling Point: 137-144° Beilstein Reference: 1901563 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00077264
: Dimethylbenzene
- :1330-20-7
- :C₆H₄(CH₃)₂
- :MFCD00077264
- :106.17 g/mol
- :14,10081
- :137…144 °C
- :< –25 °C
- :0,860
- :29 °C (84 °F)
- :Kamertemperatuur
- :215-535-7
- :1307
- :3,II
- Verpakking:Glass bottle
- VPE:1.000 mL