Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Xyleen ≥97%
Glass bottle, 1.000 mL
  L13317.AU
Xyleen ≥97%
  L13317.AU
 :  1330-20-7
Xyleen ≥97%
Xyleen ≥97%
Glass bottle, 1.000 mL
 :  1330-20-7
45,10 €
1 * 1.000 mL
Purity: 97+% CAS No.: 1330-20-7 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 29°(84°F) Refractive Index: 1.4970 Odour: Aromatic Melting Point: -25° Boiling Point: 137-144° Beilstein Reference: 1901563 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00077264
 :  Dimethylbenzene
  •  :1330-20-7
  •  :C₆H₄(CH₃)₂
  •  :MFCD00077264
  •  :106.17 g/mol
  •  :14,10081
  •  :137…144 °C
  •  :< –25 °C
  •  :0,860
  •  :29 °C (84 °F)
  •  :Kamertemperatuur
  •  :215-535-7
  •  :1307
  •  :3,II
  • Verpakking:
    Glass bottle
  • VPE:
    1.000 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.