Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Dissolution vessels, QLA
Dissolution vessels, QLA
QLA clear glass dissolution Apex vessel, serialised, 1000 ml
Catalogus # QLAAGLA9PK-01
Leverancier:   QUALITY LAB ACCESSORIES
Dissolution vessels, QLA
Catalogus # QLAAGLA9PK-01
Dissolution vessels, QLA
Dissolution vessels, QLA
QLA clear glass dissolution Apex vessel, serialised, 1000 ml
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
  • Made of high quality borosilicate glass or plastic
  • Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Specificaties
  • Omschrijving:
    QLA clear glass dissolution Apex vessel, serialised, 1000 ml
  • Seq:
    18
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.