Dissolution vessels, QLA
QLA clear glass dissolution vessel, 1000 ml
Catalogus # QLAAGLA900-EW
Leverancier: QUALITY LAB ACCESSORIES
Ga naar:
Product Details & Documents
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
- Made of high quality borosilicate glass or plastic
- Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Specificaties
- Omschrijving:QLA clear glass dissolution vessel, 1000 ml
- Seq:10
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.