Orders LinkContactUsLinkComponent
Dissolution vessels, QLA
Dissolution vessels, QLA
QLA clear glass dissolution vessel, serialised, 1000 ml
Catalogus # QLAAGLA900-01
Leverancier:   QUALITY LAB ACCESSORIES
Dissolution vessels, QLA
Catalogus # QLAAGLA900-01
Dissolution vessels, QLA
Dissolution vessels, QLA
QLA clear glass dissolution vessel, serialised, 1000 ml
Price
274,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
  • Made of high quality borosilicate glass or plastic
  • Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Specificaties
  • Omschrijving:
    QLA clear glass dissolution vessel, serialised, 1000 ml
  • Seq:
    04