Solid phase microextraction fibers
SPME fiber for autosampler use, PDMS/DVB, 24 gauge, 65 µm fiber coating
Catalogus # VARI391896314
Leverancier: VARIAN
Ga naar:
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specificaties
- Omschrijving:SPME fiber for autosampler use, PDMS/DVB, 24 gauge, 65 µm fiber coating
- Seq:04
- Cat. No.:VARI391896314
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Merlin microseal, SPME replacement seal, 23 gaugeMerlin microseal, SPME replacement seal, 23 gaugeVARI392609902Log in voor beschikbaarheid1 * 1 ST - 416,00 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- SPME fiber holder for CTC autosampler
Productinformatie
- Catalogus #BeschikbaarheidPrice
- SPME stand, 15 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- SPME inlet guide for manual injection