Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
N-Methylethylamine ≥98%

N-Methylethylamine ≥98%

Glass bottle for solids, 25 g
Catalogus # L04372.14
Ontvlambaar
Corrosief
Leverancier:   Thermo Fisher Scientific
N-Methylethylamine ≥98%
Catalogus # L04372.14
CAS-nummer:  624-78-2
N-Methylethylamine ≥98%
N-Methylethylamine ≥98%

Glass bottle for solids, 25 g

CAS-nummer:  624-78-2
Price
278,00 €
Log in voor beschikbaarheid
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 624-78-2 Molecular Formula: CH3CH2NHCH3 Formula Weight: 59.11 Storage Temperature: 2°C to 8°C Flash Point: -34°(-29°F) Refractive Index: 1.3740 Odour: Amine-like Boiling Point: 33-34° Beilstein Reference: 1730786 UN No.: 2733 MDL No.: MFCD00009030

Synoniemen:  N-Ethylmethylamine
  • CAS-nummer:624-78-2
  • Formule:C₃H₉N
  • MDL-nummer:MFCD00009030
  • Molecular Weight:59.11 g/mol
  • Kokend Pt:33…34 °C
  • Dichtheid:0,688
  • Flits Pt:<–34 °C (–29 °F)
  • EINECS:210-862-1
  • ADR:3,I
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    25 g

Specificaties