Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

N-Methylethylamine ≥98%

Glass bottle for solids, 5 g
Catalogus # L04372.06
Ontvlambaar
Corrosief
Leverancier:   Thermo Fisher Scientific
N-Methylethylamine ≥98%
Catalogus # L04372.06
CAS-nummer:  624-78-2
N-Methylethylamine ≥98%
N-Methylethylamine ≥98%

Glass bottle for solids, 5 g

CAS-nummer:  624-78-2
Price
83,80 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents

Purity: 98+% CAS No.: 624-78-2 Molecular Formula: CH3CH2NHCH3 Formula Weight: 59.11 Storage Temperature: 2°C to 8°C Flash Point: -34°(-29°F) Refractive Index: 1.3740 Odour: Amine-like Boiling Point: 33-34° Beilstein Reference: 1730786 UN No.: 2733 MDL No.: MFCD00009030

Synoniemen:  N-Ethylmethylamine
  • CAS-nummer:624-78-2
  • Formule:C₃H₉N
  • MDL-nummer:MFCD00009030
  • Molecular Weight:59.11 g/mol
  • Kokend Pt:33…34 °C
  • Dichtheid:0,688
  • Flits Pt:<–34 °C (–29 °F)
  • EINECS:210-862-1
  • ADR:3,I
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    5 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.