Transfer pipettes, Samco™ Extra Long
5,6 ml, 25, 2,3 ml, 5,0 mm, 225 mm, +, Individually wrapped
Catalogus # SAMC262-1S
Leverancier: SAMCO
Transfer pipettes, Samco™ Extra Long
Catalogus # SAMC262-1S
Leverancier: SAMCO
Ga naar:
Product Details & Documents
These extra long PE pipettes are ideal for safe and easy sampling from large containers.
- Excellent for tissue culture applications
- General purpose pipettes are ideal for routine sampling of serum and plasma
- Well suited for blood culture and urine analysis
Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination.
Specificaties
- Ø uitw.:5,0 mm
- Verpakt:Individually wrapped
- Steriel:+
- Capaciteit:5,6 ml
- Lengte:225 mm
- Druppels per ml:25
- Aanzuigvolume:2,3 ml
- Milieu certificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Lage impact op de fabricage
- Cat. No.:SAMC262-1S
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



