Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
-
-
Apparaat arsenicumbepaling
Catalogus # 201-8003
Leverancier:   SciLabware
-
Catalogus # 201-8003
Leverancier:  SciLabware
-
-
Apparaat arsenicumbepaling
Price
226,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass arsenic determination apparatus designed for the estimation of arsenic and antimony.
  • Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 5380A
  • Complete assembly (all parts single available)
Specificaties
  • Omschrijving:
    Apparaat arsenicumbepaling
  • Seq:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.