-
Apparaat arsenicumbepaling
Catalogus # 201-8003
Leverancier: SciLabware
Ga naar:
Product Details & Documents
PYREX®, borosilicate glass arsenic determination apparatus designed for the estimation of arsenic and antimony.
- Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 5380A
- Complete assembly (all parts single available)
Specificaties
- Omschrijving:Apparaat arsenicumbepaling
- Seq:01