Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Unisette™ M506
abrikoos
Catalogus # 720-1575
Leverancier:   Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M506
Catalogus # 720-1575
Leverancier:  Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M506
Biopsy processing/embedding cassettes, Unisette™ M506
abrikoos
Price
267,00 €
Log in voor beschikbaarheid
1 * 1.500 ST
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Integral lid
  • Front face inclined at an angle of 45° so that the cassette can be used with certain types of cassette labelling devices
  • Available in 11 non cytotoxic, non metallic colours and three fluorescent colours
 : Each case contains three boxes of 500 cassettes each.
Specificaties
  • Kleur:
    abrikoos
  • Seq:
    06