Microtubes, with Thread
Micro tubes without screw cap, without skirted base, 1,5 ml, 5000/case
Catalogus # SARS72.607
Marketsource
Leverancier: SARSTEDT
Ga naar:
Product Details & Documents
PP, translucent.
- Safe screw cap with O-ring
- Sterile or non sterile
- Autoclavable
For transport, storage and sample processing in the laboratory.
Specificaties
- Verpakt:5000/case
- Capaciteit:1,5 ml
- Omschrijving:Micro tubes without screw cap, without skirted base