Cell scrapers
Celschraper met twee bladposities, 250 mm
Catalogus # SARS83.3951
Marketsource
Leverancier: SARSTEDT
Ga naar:
Product Details & Documents
Cell scraper with soft, flexible blade for gentle removal of monolayer cultures, minimising cell damage.
- Easy and complete separation of monolayer due to extra thin and flexible blade
- When downward pressure is applied to the handle the blade can remove all cells in its path in one pass
- Sterilised by gamma irradiation, individually wrapped
Specificaties
- Omschrijving:Celschraper met twee bladposities
- Seq:01
- Totale lengte:250 mm