Orders LinkContactUsLinkComponent
Cell scrapers
Cell scrapers
Celschraper met twee bladposities, 250 mm
Catalogus # SARS83.3951
Marketsource
Leverancier:   SARSTEDT
Cell scrapers
Catalogus # SARS83.3951
Leverancier:  SARSTEDT
Marketsource
Cell scrapers
Celschraper met twee bladposities, 250 mm
Price
171,54 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
Cell scraper with soft, flexible blade for gentle removal of monolayer cultures, minimising cell damage.
  • Easy and complete separation of monolayer due to extra thin and flexible blade
  • When downward pressure is applied to the handle the blade can remove all cells in its path in one pass
  • Sterilised by gamma irradiation, individually wrapped
Specificaties
  • Omschrijving:
    Celschraper met twee bladposities
  • Seq:
    01
  • Totale lengte:
    250 mm