Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Ethyl bromopyruvate ≥75%, Technical Grade

Ethyl bromopyruvate ≥75%, Technical Grade

Glass bottle, 25 g
Catalogus # L00582.14
Corrosief
Leverancier:   Thermo Fisher Scientific
Ethyl bromopyruvate ≥75%, Technical Grade
Catalogus # L00582.14
CAS-nummer:  70-23-5
Ethyl bromopyruvate ≥75%, Technical Grade
Ethyl bromopyruvate ≥75%, Technical Grade

Glass bottle, 25 g

CAS-nummer:  70-23-5
Price
60,50 €
Log in voor beschikbaarheid
1 * 25 g
Product Details & Documents

Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide

Purity: 85% CAS No.: 70-23-5 Molecular Formula: C5H7BrO3 Formula Weight: 195.02 Flash Point: 98°(208°F) Refractive Index: 1.4690 Boiling Point: 98-100°/10mm Beilstein Reference: 1760158 UN No.: 3265 MDL No.: MFCD00000204

Synoniemen:  Bromopyruvic acid ethyl ester, Ethyl 3-bromo-2-oxopropanoate, 3-Bromo-2-oxopropionic acid ethyl ester
  • CAS-nummer:70-23-5
  • Formule:BrH₂CC(O)COOCH₂CH₃
  • MDL-nummer:MFCD00000204
  • Molecular Weight:195.01 g/mol
  • Kokend Pt:98…100 °C (10 mmHg)
  • Dichtheid:1,555
  • Flits Pt:98 °C (208 °F)
  • Temperatuur bij opslag:Koelkast
  • EINECS:200-729-6
  • UN:3265
  • ADR:8,III
  •  :Density: 1.55 g/cm³ (20 °C)
Specificaties

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    25 g

Specificaties