Accessories for comparators
Bromine, 3/53C, 0,5 - 6,0 mg/l
N° de catalogue LOVI235330
Fournisseur: Lovibond
Aller à :
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Spécifications
- Meetbereik:0,5 - 6,0 mg/l
- Seq:06
- Code:3/53C
- Testmethode:Bromine