Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Accessories for comparators
Accessories for comparators
Bromine, 3/53C, 0,5 - 6,0 mg/l
N° de catalogue LOVI235330
Fournisseur:   Lovibond
Accessories for comparators
N° de catalogue LOVI235330
Fournisseur:  Lovibond
Accessories for comparators
Accessories for comparators
Bromine, 3/53C, 0,5 - 6,0 mg/l
Prix
263,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Spécifications
  • Meetbereik:
    0,5 - 6,0 mg/l
  • Seq:
    06
  • Code:
    3/53C
  • Testmethode:
    Bromine
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.