Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Accessories for comparators
Accessories for comparators
QAC (Quaternary Ammonia Compounds), 3/118, 2 - 20 mg/l
Catalogus # LOVI230120
Leverancier:   Lovibond
Accessories for comparators
Catalogus # LOVI230120
Leverancier:  Lovibond
Accessories for comparators
Accessories for comparators
QAC (Quaternary Ammonia Compounds), 3/118, 2 - 20 mg/l
Price
283,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specificaties
  • Meetbereik:
    2 - 20 mg/l
  • Seq:
    71
  • Code:
    3/118
  • Testmethode:
    QAC (Quaternary Ammonia Compounds)
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.