Accessories for comparators
pH, 2/1F, 4,8 - 6,4 pH
Catalogus # LOVI221090
Leverancier: Lovibond
Ga naar:
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Specificaties
- Meetbereik:4,8 - 6,4 pH
- Seq:56
- Code:2/1F
- Testmethode:pH