Pouring rings
Pouring ring, GL 45, helder
Catalogus # 215-3138
Leverancier: SciLabware
Ga naar:
Product Details & Documents
Pouring rings, made of Polypropylene, for laboratory/media bottles.
- Ensures drip-free pouring
- Very good chemical resistance
- Autoclavable up to 140 °C
Specificaties
- Schroefdraad:GL 45
- Kleur:helder
- Omschrijving:Pouring ring
- Seq:02