Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
-
-
Absorption tube with cone size 24/29 for arsenic determination apparatus
Catalogus # BIBBAD3
Leverancier:   SciLabware
-
Catalogus # BIBBAD3
Leverancier:  SciLabware
-
-
Absorption tube with cone size 24/29 for arsenic determination apparatus
Price
207,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass arsenic determination apparatus designed for the estimation of arsenic and antimony.
  • Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 5380A
  • Complete assembly (all parts single available)
Specificaties
  • Omschrijving:
    Absorption tube with cone size 24/29 for arsenic determination apparatus
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.