-
Absorption tube with cone size 24/29 for arsenic determination apparatus
Catalogus # BIBBAD3
Leverancier: SciLabware
Ga naar:
Product Details & Documents
PYREX®, borosilicate glass arsenic determination apparatus designed for the estimation of arsenic and antimony.
- Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 5380A
- Complete assembly (all parts single available)
Specificaties
- Omschrijving:Absorption tube with cone size 24/29 for arsenic determination apparatus
- Seq:02