Orders LinkContactUsLinkComponent
Flessendragers
Maximaal 6 flessen, 74 mm
  215-3277
 :   SciLabware
Flessendragers
  215-3277
Flessendragers
Flessendragers
Maximaal 6 flessen, 74 mm
132,00 €
1 * 1 ST
Staal, PA-coating.
These bottle carriers provide a high resistance to heat and chemicals. They are designed for safe and reliable transport of bottles.
  • Voor:
    Maximaal 6 flessen
  • Max. Ø recipiënt:
    74 mm