Flessendragers
Maximaal 6 flessen, 74 mm
215-3277
Staal, PA-coating.
These bottle carriers provide a high resistance to heat and chemicals. They are designed for safe and reliable transport of bottles.
- Voor:Maximaal 6 flessen
- Max. Ø recipiënt:74 mm