Flessendragers
Maximaal 6 flessen, 60 mm
Catalogus # 215-3276
Leverancier: SciLabware
Ga naar:
Product Details & Documents
Staal, PA-coating.
These bottle carriers provide a high resistance to heat and chemicals. They are designed for safe and reliable transport of bottles.
Specificaties
- Voor:Maximaal 6 flessen
- Max. Ø recipiënt:60 mm