ContactUsLinkComponent
CELLMASTER# roller bottles 2X, van heldere, breukvaste kunststof (PET) non pyrogenic , DNase/RNase free, oppervl. 1700 cm, vulv olume: 2500 ml, graduering 50-1000 ml, lengte 275 mm, diam. 122 mm, per 5 onderverpakt STERIEL 1 * 20 ST
  392-0067
CELLMASTER# roller bottles 2X, van heldere, breukvaste kunststof (PET) non pyrogenic , DNase/RNase free, oppervl. 1700 cm, vulv olume: 2500 ml, graduering 50-1000 ml, lengte 275 mm, diam. 122 mm, per 5 onderverpakt STERIEL 1 * 20 ST
  392-0067
Frequently Bought Together