Celtellers, Nageotte
Celteller, donkere bekleding, met klemmen Nageotte, 0,5 mm
Catalogus # MARI0640511
Leverancier: MARIENFELD
Ga naar:
Product Details & Documents
These counting chambers are designed for leucocytes and erythrocytes in in vitro applications.
: IVD directive 98/79/EC.
Specificaties
- Kamerdiepte:0,5 mm
- Omschrijving:Celteller, donkere bekleding, met klemmen Nageotte
- Seq:002