Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Celtellers, Neubauer
Celtellers, Neubauer
Counting chamber, dark-lined, Neubauer, 0,10 mm
Catalogus # MARI0640110
Leverancier:   MARIENFELD
Celtellers, Neubauer
Catalogus # MARI0640110
Leverancier:  MARIENFELD
Celtellers, Neubauer
Counting chamber, dark-lined, Neubauer, 0,10 mm
Price
51,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These counting chambers are designed for counting leucocytes and erythrocytes.
  • Double net ruling
  • Two counting grids with one division
 : IVD 98/79/EC
 : Supplied with two cover glasses.
Documents
Specificaties
  • Kamerdiepte:
    0,10 mm
  • Omschrijving:
    Counting chamber, dark-lined, Neubauer
  • Seq:
    03