Orders LinkContactUsLinkComponent
Temperature control jackets for viscosity cups
Temperature control jackets for viscosity cups
Temperature control jacket with tripod for ISO and AFNOR cups
N° de catalogue SIMEVF2068
Fournisseur:   Industrial Physics
Temperature control jackets for viscosity cups
N° de catalogue SIMEVF2068
Fournisseur:  Industrial Physics
Temperature control jackets for viscosity cups
Temperature control jackets for viscosity cups
Temperature control jacket with tripod for ISO and AFNOR cups
Prix
621,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The temperature control jacket for viscosity cups is a double jacketed casing of anodised aluminium with built-in spirit level to bring the viscosity beaker to the required temperature with cooled and heated liquids.
  • Adjustable feet for easy leveling
  • Built-in spirit level to set jacket exactly horizontal
  • Glass plate to prevent dripping before measurement
  • Elbow-type quick release couplings
  • Titanium anodised aluminium
Livraison: Supplied with jacket with built-in spirit level, glass plate and tube connection.
Services: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Documents
Spécifications
  • Omschrijving:
    Temperature control jacket with tripod for ISO and AFNOR cups
  • Seq:
    02
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.