Cover glasses
24×40 mm, #1, Borosilicaatglas
Supplier: MARIENFELD
Jump to:
Product Details & Documents
These cover glasses are suitable for fluorescence microscopy and in vitro diagnostic (IVD) applications.
- Long storage, non corroding
- Made of clear borosilicate D 263™ M glass, Hydrolytic Class I
- Do not stick together
- Excellent flat, smooth surfaces
Certifications: IVD 98/79/EC.
Packaging: In plastic boxes with hinged lid – 1000 pieces per box.
Documents
Certificates
Search by Lot NumberSpecifications
- Aantal:#1
- l×b:24×40 mm
- Materiaal:Borosilicaatglas