2-(4-Morpholinyl)phenol
Glass bottle for solids, 1 g
Catalogus # MAYBS08696DA
Marketsource
Leverancier: Thermo Scientific
2-(4-Morpholinyl)phenol
Catalogus # MAYBS08696DA
Leverancier: Thermo Scientific
Marketsource
CAS-nummer: 41536-44-1
Ga naar:
Product Details & Documents
Synoniemen: 2-Morpholinophenol
- CAS-nummer:41536-44-1
- Formule:C₁₀H₁₃NO₂
- MDL-nummer:MFCD00051674
- Molecular Weight:179.215 g/mol
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:1 g



