Nitrofen
Glass bottle for solids, 1 g
Catalogus # MAYBRDP00053DA
Marketsource
Leverancier: Thermo Scientific
Ga naar:
Product Details & Documents
Synoniemen: 2,4-Dichloro-1-(4-nitrophenoxy)benzene
- CAS-nummer:1836-75-5
- Formule:C₁₂H₇Cl₂NO₃
- MDL-nummer:MFCD00128026
- Molecular Weight:284.1 g/mol
- EINECS:217-406-0
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:1 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



