Kroezen
30 ml, 40×40 mm, 0,5 mm
Catalogus # BOCH8104
Leverancier: BOCHEM
Ga naar:
Product Details & Documents
These crucibles and lids are made of nickel or stainless steel 18/10.
: Lids need to be ordered separately.
Specificaties
- Ø×h:40×40 mm
- Capaciteit:30 ml
- Seq:07
- Dikte:0,5 mm