PH-combinatie-elektroden
Sensolyt® DW*, Sensolyt® DW, without arming, to be installed into flow vessels, 0 - 14, Drinking water
Catalogus # WTWO109103
Leverancier: WTW ONLINE
Ga naar:
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specificaties
- Voor:Drinking water
- Type:Sensolyt® DW, without arming, to be installed into flow vessels
- Omschrijving:Sensolyt® DW*
- Seq:33
- pH:0 - 14
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.