Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Agarose

Agarose

Glass bottle for solids, 250 g
Catalogus # SERA11381.03
Marketsource
Leverancier:   Serva
Agarose
Catalogus # SERA11381.03
Leverancier:  Serva
Marketsource
CAS-nummer:  9012-36-6
Agarose
Agarose

Glass bottle for solids, 250 g

CAS-nummer:  9012-36-6
Price
350,00 €
Log in voor beschikbaarheid
1 * 250 g
Product Details & Documents
  • CAS-nummer:9012-36-6
  • MDL-nummer:MFCD00081294
  • Smelten Pt:60…90 °C
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:232-731-8
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    250 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.